Methyl-3-amino-4-brombenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-amino-4-bromobenzoate

AS32383G1 AS32383G5
ACTIAS32383G1EA 0 CHF
ACTIAS32383G1 ACTIAS32383G5
Methyl-3-amino-4-brombenzoat
Methyl-3-amino-4-brombenzoat
Formel: C₈H₈BrNO₂
MDL Number: MFCD00553090
CAS-Nummer: 46064-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR