Methyl-2-amino-3-brombenzoat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Methyl 2-amino-3-bromobenzoate
AS34453G1
AS34453G5
ACTIAS34453G1EA
0
CHF
ACTIAS34453G1
ACTIAS34453G5
Methyl-2-amino-3-brombenzoat
Methyl-2-amino-3-brombenzoat
Formel:
C₈H₈BrNO₂ MW: 230,0586 g/mol |
MDL Number:
MFCD11042862 CAS-Nummer: 104670-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...