(2-Brom-5-fluorphenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-5-fluorophenylacetic acid

AS35307G1 AS35307G5
ACTIAS35307G1EA 0 CHF
ACTIAS35307G1 ACTIAS35307G5
(2-Brom-5-fluorphenyl)essigsäure
(2-Brom-5-fluorphenyl)essigsäure
Formel: C₈H₆BrFO₂
MW: 233,03 g/mol
MDL Number: MFCD03094440
CAS-Nummer: 739336-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR