6-Hydroxynicotinonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(4-Formylphenyl)isonicotinonitrile, 6-Hydroxypyridine-3-carbonitrile, 5-Cyano-2-hydroxypyridine

AS35858G1 AS35858G5
ACTIAS35858G1EA 0 CHF
ACTIAS35858G1 ACTIAS35858G5
6-Hydroxynicotinonitril
6-Hydroxynicotinonitril
Formel: C₆H₄N₂O
MW: 120,109 g/mol
MDL Number: MFCD06656489
CAS-Nummer: 94805-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR