4-Amino-N-methylbenzamid

Lieferant: ACTIVATE SCIENTIFIC

AS21925G100 AS21925G25
ACTIAS21925G100EA 138 CHF
ACTIAS21925G100 ACTIAS21925G25
4-Amino-N-methylbenzamid
4-Amino-N-methylbenzamid
Formel: C₈H₁₀N₂O
MW: 150,1778 g/mol
MDL Number: MFCD00665792
CAS-Nummer: 6274-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR