3-Amino-N-methylbenzamid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Aminobenzoylmethylamide

AS36405G1 AS36405G5
ACTIAS36405G1EA 0 CHF
ACTIAS36405G1 ACTIAS36405G5
3-Amino-N-methylbenzamid
3-Amino-N-methylbenzamid
Formel: C₈H₁₀N₂O
MW: 150,18 g/mol
MDL Number: MFCD00070630
CAS-Nummer: 25900-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR