5-Methoxy-2-nitrobenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS31724G1 AS31724G5
ACTIAS31724G1EA 0 CHF
ACTIAS31724G1 ACTIAS31724G5
5-Methoxy-2-nitrobenzonitril
5-Methoxy-2-nitrobenzonitril
Formel: C₈H₆N₂O₃
MDL Number: MFCD09056823
CAS-Nummer: 38469-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR