2-Brom-4-fluor-5-nitrophenol

Lieferant: ACTIVATE SCIENTIFIC

AS22694G1 AS22694G5
ACTIAS22694G1EA 0 CHF
ACTIAS22694G1 ACTIAS22694G5
2-Brom-4-fluor-5-nitrophenol
2-Brom-4-fluor-5-nitrophenol
Formel: C₆H₃BrFNO₃
MDL Number: MFCD08460104
CAS-Nummer: 84478-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR