3-(Trifluormethyl)isonicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-(Trifluoromethyl)pyridine-4-carboxylic acid

AS32424G1 AS32424G5
ACTIAS32424G1EA 0 CHF
ACTIAS32424G1 ACTIAS32424G5
3-(Trifluormethyl)isonicotinsäure
3-(Trifluormethyl)isonicotinsäure
Formel: C₇H₄F₃NO₂
MW: 191,11 g/mol
MDL Number: MFCD08234940
CAS-Nummer: 590371-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR