4-Methoxyisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS34007G1 AS34007G5
ACTIAS34007G1EA 0 CHF
ACTIAS34007G1 ACTIAS34007G5
4-Methoxyisochinolin
4-Methoxyisoquinoline
Formel: C₁₀H₉NO
MW: 159,1846 g/mol
MDL Number: MFCD01646265
CAS-Nummer: 36034-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR