6-Fluorisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS36110G1 AS36110G5
ACTIAS36110G1EA 0 CHF
ACTIAS36110G1 ACTIAS36110G5
6-Fluorisochinolin
6-Fluoroisoquinoline
Formel: C₉H₆FN
MW: 147,1491 g/mol
MDL Number: MFCD11226829
CAS-Nummer: 1075-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR