1,6-Dichlorisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS36596G5 AS36596G1
ACTIAS36596G5EA 0 CHF
ACTIAS36596G5 ACTIAS36596G1
1,6-Dichlorisochinolin
1,6-Dichloroisoquinoline
Formel: C₉H₅Cl₂N
MW: 198.0500 g/mol
MDL Number: MFCD06738660
CAS-Nummer: 630421-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR