1,4-Dichlorisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS32269G1 AS32269G5
ACTIAS32269G1EA 0 CHF
ACTIAS32269G1 ACTIAS32269G5
1,4-Dichlorisochinolin
1,4-Dichloroisoquinoline
Formel: C₉H₅Cl₂N
MW: 198.0500 g/mol
MDL Number: MFCD05664199
CAS-Nummer: 15298-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR