6-Brom-1,3-dichlorisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS51078G1 AS51078G5
ACTIAS51078G1EA 0 CHF
ACTIAS51078G1 ACTIAS51078G5
6-Brom-1,3-dichlorisochinolin
6-Bromo-1,3-dichloroisoquinoline
Formel: C₉H₄BrCl₂N
MW: 276.9448 g/mol
MDL Number: MFCD09258639
CAS-Nummer: 552331-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR