5-Brom-1-chlorisochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS34375G1 AS34375G5
ACTIAS34375G1EA 0 CHF
ACTIAS34375G1 ACTIAS34375G5
5-Brom-1-chlorisochinolin
5-Bromo-1-chloroisoquinoline
Formel: C₉H₅BrClN
MW: 242,4997 g/mol
MDL Number: MFCD08703278
CAS-Nummer: 34551-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR