4-Isochinolinol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Isoquinolin-4-ol, 4-Hydroxyisoquinoline

AS11009G1 AS11009G5
ACTIAS11009G1EA 0 CHF
ACTIAS11009G1 ACTIAS11009G5
4-Isochinolinol
4-Isochinolinol
Formel: C₉H₇NO
MW: 145,1580 g/mol
MDL Number: MFCD00234514
CAS-Nummer: 3336-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR