8-Isochinolincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Isoquinoline-8-carboxylic acid

AS1314G1 AS1314G5
ACTIAS1314G1EA 0 CHF
ACTIAS1314G1 ACTIAS1314G5
8-Isochinolincarbonsäure
8-Isoquinolinecarboxylic acid
Formel: C₁₀H₇NO₂
MW: 173,1681 g/mol
MDL Number: MFCD06738935
CAS-Nummer: 61563-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR