6-Isochinolincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Isoquinolinecarboxylic Acid

AS1312G1 AS1312G5
ACTIAS1312G1EA 0 CHF
ACTIAS1312G1 ACTIAS1312G5
6-Isochinolincarbonsäure
6-Isoquinolinecarboxylic acid
Formel: C₁₀H₇NO₂
MW: 173,1681 g/mol
MDL Number: MFCD09910323
CAS-Nummer: 106778-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR