5-Isochinolincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Isoquinoline-5-carboxylic acid

AS1311G1 AS1311G5
ACTIAS1311G1EA 0 CHF
ACTIAS1311G1 ACTIAS1311G5
5-Isochinolincarbonsäure
5-Isoquinolinecarboxylic acid
Formel: C₁₀H₇NO₂
MW: 173,17 g/mol
MDL Number: MFCD03788744
CAS-Nummer: 27810-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR