4-Isochinolincarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Isoquinoline-4-carboxylic acid

AS1310G1 AS1310G5
ACTIAS1310G1EA 0 CHF
ACTIAS1310G1 ACTIAS1310G5
4-Isochinolincarbonsäure
4-Isoquinolinecarboxylic acid
Formel: C₁₀H₇NO₂
MW: 173,1681 g/mol
MDL Number: MFCD00094344
CAS-Nummer: 7159-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR