3-Bromisochinolin
Lieferant: ACTIVATE SCIENTIFIC
AS1287G1
AS1287G5
ACTIAS1287G1EA
0
CHF
ACTIAS1287G1
ACTIAS1287G5
3-Bromisochinolin
3-Bromoisoquinoline
Formel:
C₉H₆BrN MW: 208,0600 g/mol |
MDL Number:
MFCD00234479 CAS-Nummer: 34784-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...