Methyl-2-hydroxyisonicotinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 1,2-dihydro-2-oxopyridine-4-carboxylate

AS31926G1 AS31926G5
ACTIAS31926G1EA 0 CHF
ACTIAS31926G1 ACTIAS31926G5
Methyl-2-hydroxyisonicotinat
Methyl-2-hydroxyisonicotinat
Formel: C₇H₇NO₃
MW: 153,1400 g/mol
MDL Number: MFCD09258780
CAS-Nummer: 89937-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR