Methyl-4-fluor-2-methylbenzoat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Methyl 4-fluoro-2-methylbenzoate
AS35360G1
AS35360G5
ACTIAS35360G1EA
0
CHF
ACTIAS35360G1
ACTIAS35360G5
Methyl-4-fluor-2-methylbenzoat
Methyl-4-fluor-2-methylbenzoat
Formel:
C₉H₉FO₂ MW: 168.16 g/mol |
MDL Number:
MFCD06203790 CAS-Nummer: 174403-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...