Methyl-3-fluor-4-methylbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-fluoro-4-methylbenzoate

AS32636G1 AS32636G5
ACTIAS32636G1EA 0 CHF
ACTIAS32636G1 ACTIAS32636G5
Methyl-3-fluor-4-methylbenzoat
Methyl-3-fluor-4-methylbenzoat
Formel: C₉H₉FO₂
MW: 168.1650 g/mol
MDL Number: MFCD06203786
CAS-Nummer: 87808-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR