Methyl-2-fluor-5-methylbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-fluoro-5-methylbenzoate

AS24914G100 AS24914G25 AS24914G5
ACTIAS24914G100EA 490 CHF
ACTIAS24914G100 ACTIAS24914G25 ACTIAS24914G5
Methyl-2-fluor-5-methylbenzoat
Methyl-2-fluor-5-methylbenzoat
Formel: C₉H₉FO₂
MW: 168,1700 g/mol
MDL Number: MFCD06203924
CAS-Nummer: 2967-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR