Methyl-3-brom-2-methylbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 3-bromo-2-methylbenzoate

AS23229G25 AS23229G5
ACTIAS23229G25EA 37 CHF
ACTIAS23229G25 ACTIAS23229G5
Methyl-3-brom-2-methylbenzoat
Methyl-3-brom-2-methylbenzoat
Formel: C₉H₉BrO₂
MW: 229,0706 g/mol
MDL Number: MFCD09031772
CAS-Nummer: 99548-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR