Methyl-2-brom-6-methylbenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 2-bromo-6-methylbenzoate

AS33561G1 AS33561G5
ACTIAS33561G1EA 0 CHF
ACTIAS33561G1 ACTIAS33561G5
Methyl-2-brom-6-methylbenzoat
Methyl-2-brom-6-methylbenzoat
Formel: C₉H₉BrO₂
MW: 229,0700 g/mol
MDL Number: MFCD09260938
CAS-Nummer: 99548-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR