5-Chinolylmethylether

Lieferant: ACTIVATE SCIENTIFIC

AS40523G1 AS40523G5
ACTIAS40523G1EA 0 CHF
ACTIAS40523G1 ACTIAS40523G5
5-Chinolylmethylether
5-Chinolylmethylether
Formel: C₁₀H₉NO
MDL Number: MFCD02752664
CAS-Nummer: 6931-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR