3-Chinolylmethylether

Lieferant: ACTIVATE SCIENTIFIC

AS32609G1 AS32609G5
ACTIAS32609G1EA 0 CHF
ACTIAS32609G1 ACTIAS32609G5
3-Chinolylmethylether
3-Chinolylmethylether
Formel: C₁₀H₉NO
MDL Number: MFCD16619425
CAS-Nummer: 6931-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR