2,3-Dibromchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS32668G1 AS32668G5
ACTIAS32668G1EA 0 CHF
ACTIAS32668G1 ACTIAS32668G5
2,3-Dibromchinolin
2,3-Dibromoquinoline
Formel: C₉H₅Br₂N
MDL Number: MFCD00234472
CAS-Nummer: 13721-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR