2-Pyridinboronsäuredimethylester

Lieferant: ACTIVATE SCIENTIFIC

AS51531G1 AS51531G5
ACTIAS51531G1EA 0 CHF
ACTIAS51531G1 ACTIAS51531G5
2-Pyridinboronsäuredimethylester
2-Pyridineboronic acid dimethyl ester
Formel: C₇H₁₀BNO₂
MW: 150.97 g/mol
MDL Number: MFCD03412109
CAS-Nummer: 136805-54-4

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR