(2-Fluor-4-nitrophenyl)essigsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(2-Fluoro-4-nitrophenyl)acetic acid

AS21951G1 AS21951G5
ACTIAS21951G1EA 0 CHF
ACTIAS21951G1 ACTIAS21951G5
(2-Fluor-4-nitrophenyl)essigsäure
(2-Fluor-4-nitrophenyl)essigsäure
Formel: C₈H₆FNO₄
MDL Number: MFCD11041422
CAS-Nummer: 315228-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR