3-Amino-5-fluorbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS33940G1 AS33940G5
ACTIAS33940G1EA 0 CHF
ACTIAS33940G1 ACTIAS33940G5
3-Amino-5-fluorbenzoesäure
3-Amino-5-fluorobenzoic acid
Formel: C₇H₆FNO₂
MW: 155,13 g/mol
MDL Number: MFCD07779526
CAS-Nummer: 786616-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR