4-Brom-2-fluor-5-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS33922G1 AS33922G5
ACTIAS33922G1EA 0 CHF
ACTIAS33922G1 ACTIAS33922G5
4-Brom-2-fluor-5-methylbenzonitril
4-Brom-2-fluor-5-methylbenzonitril
Formel: C₈H₅BrFN
MW: 214.0300 g/mol
MDL Number: MFCD07782073
CAS-Nummer: 916792-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR