2-Brom-4-fluor-6-methylbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS35086G1 AS35086G10 AS35086G5
ACTIAS35086G1EA 0 CHF
ACTIAS35086G1 ACTIAS35086G10 ACTIAS35086G5
2-Brom-4-fluor-6-methylbenzonitril
2-Brom-4-fluor-6-methylbenzonitril
Formel: C₈H₅BrFN
MW: 214.0300 g/mol
MDL Number: MFCD07782071
CAS-Nummer: 916792-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR