4-Chlor-2,5-difluorbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS32669G1 AS32669G5
ACTIAS32669G1EA 0 CHF
ACTIAS32669G1 ACTIAS32669G5
4-Chlor-2,5-difluorbenzaldehyd
4-Chloro-2,5-difluorobenzaldehyde
Formel: C₇H₃ClF₂O
MDL Number: MFCD07782051
CAS-Nummer: 879093-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR