2-Chlor-3-fluorbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS31520G1 AS31520G5
ACTIAS31520G1EA 0 CHF
ACTIAS31520G1 ACTIAS31520G5
2-Chlor-3-fluorbenzaldehyd
2-Chloro-3-fluorobenzaldehyde
Formel: C₇H₄ClFO
MDL Number: MFCD07782053
CAS-Nummer: 96516-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR