2,4-Dichlor-6-fluorbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS13303G1 AS13303G5
ACTIAS13303G1EA 0 CHF
ACTIAS13303G1 ACTIAS13303G5
2,4-Dichlor-6-fluorbenzaldehyd
2,4-Dichloro-6-fluorobenzaldehyde
Formel: C₇H₃Cl₂FO
MDL Number: MFCD12404935
CAS-Nummer: 681435-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR