3-Amino-5-chlorbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS23384G1 AS23384G5
ACTIAS23384G1EA 0 CHF
ACTIAS23384G1 ACTIAS23384G5
3-Amino-5-chlorbenzoesäure
3-Amino-5-chlorobenzoic acid
Formel: C₇H₆ClNO₂
MW: 171,5810 g/mol
MDL Number: MFCD02183894
CAS-Nummer: 21961-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR