4-Chlor-2,6-dinitrobenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS23898G1 AS23898G5
ACTIAS23898G1EA 747 CHF
ACTIAS23898G1 ACTIAS23898G5
4-Chlor-2,6-dinitrobenzonitril
4-Chlor-2,6-dinitrobenzonitril
Formel: C₇H₂ClN₃O₄
MDL Number: MFCD18651708
CAS-Nummer: 1272756-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR