5-Methoxy-2-nitrobenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30702G1 AS30702G5
ACTIAS30702G1EA 0 CHF
ACTIAS30702G1 ACTIAS30702G5
5-Methoxy-2-nitrobenzaldehyd
5-Methoxy-2-nitrobenzaldehyd
Formel: C₈H₇NO₄
MW: 181,1500 g/mol
MDL Number: MFCD00456498
CAS-Nummer: 20357-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR