1-Brom-3-fluor-5-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

AS32526G1 AS32526G5
ACTIAS32526G1EA 0 CHF
ACTIAS32526G1 ACTIAS32526G5
1-Brom-3-fluor-5-nitrobenzol
1-Bromo-3-fluoro-5-nitrobenzene
Formel: C₆H₃BrFNO₂
MW: 220.00 g/mol
MDL Number: MFCD09955454
CAS-Nummer: 7087-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR