5-Brom-1,3-difluor-2-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3,5-Difluoro-4-nitrobromobenzene

AS32261G1 AS32261G5
ACTIAS32261G1EA 0 CHF
ACTIAS32261G1 ACTIAS32261G5
5-Brom-1,3-difluor-2-nitrobenzol
5-Brom-1,3-difluor-2-nitrobenzol
Formel: C₆H₂BrF₂NO₂
MW: 237,9864 g/mol
MDL Number: MFCD12025434
CAS-Nummer: 147808-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR