1-Brom-3,5-difluor-2-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

AS32499G1 AS32499G5
ACTIAS32499G1EA 0 CHF
ACTIAS32499G1 ACTIAS32499G5
1-Brom-3,5-difluor-2-nitrobenzol
1-Bromo-3,5-difluoro-2-nitrobenzene
Formel: C₆H₂BrF₂NO₂
MDL Number: MFCD04112501
CAS-Nummer: 884494-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR