1-Brom-2,3-difluor-4-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,3-Difluoro-4-bromonitrobenzene

AS33672G1 AS33672G5
ACTIAS33672G1EA 0 CHF
ACTIAS33672G1 ACTIAS33672G5
1-Brom-2,3-difluor-4-nitrobenzol
1-Bromo-2,3-difluoro-4-nitrobenzene
Formel: C₆H₂BrF₂NO₂
MW: 237,9900 g/mol
MDL Number: MFCD09839215
CAS-Nummer: 1003708-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR