1-Brom-4-chlor-3-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromo-1-chloro-2-nitrobenzene, 5-Bromo-2-chloronitrobenzene

AS32278G1 AS32278G5
ACTIAS32278G1EA 0 CHF
ACTIAS32278G1 ACTIAS32278G5
1-Brom-4-chlor-3-nitrobenzol
1-Bromo-4-chloro-3-nitrobenzene
Formel: C₆H₃BrClNO₂
MW: 236.4500 g/mol
MDL Number: MFCD05863216
CAS-Nummer: 16588-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR