1-Brom-5-chlor-2-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

AS33867G1 AS33867G5
ACTIAS33867G1EA 0 CHF
ACTIAS33867G1 ACTIAS33867G5
1-Brom-5-chlor-2-nitrobenzol
1-Brom-5-chlor-2-nitrobenzol
Formel: C₆H₃BrClNO₂
MDL Number: MFCD08437614
CAS-Nummer: 63860-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR