1-Brom-2-chlor-3-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

AS33807G1 AS33807G5
ACTIAS33807G1EA 0 CHF
ACTIAS33807G1 ACTIAS33807G5
1-Brom-2-chlor-3-nitrobenzol
1-Bromo-2-chloro-3-nitrobenzene
Formel: C₆H₃BrClNO₂
MW: 236.4505 g/mol
MDL Number: MFCD08277301
CAS-Nummer: 3970-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR