3-Brom-o-anisaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS30568G1 AS30568G5
ACTIAS30568G1EA 0 CHF
ACTIAS30568G1 ACTIAS30568G5
3-Brom-o-anisaldehyd
3-Brom-o-anisaldehyd
Formel: C₈H₇BrO₂
MDL Number: MFCD09834811
CAS-Nummer: 88275-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR