Methyl-4-amino-3-methoxybenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-amino-3-methoxybenzoate

AS30754G1 AS30754G5
ACTIAS30754G1EA 0 CHF
ACTIAS30754G1 ACTIAS30754G5
Methyl-4-amino-3-methoxybenzoat
Methyl-4-amino-3-methoxybenzoat
Formel: C₉H₁₁NO₃
MW: 181.1900 g/mol
MDL Number: MFCD00017203
CAS-Nummer: 41608-64-4
EINECS: 255-456-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR